Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3,5-dimethylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,5-Dimethylanthranilic acid

L07003-06 L07003-03 L07003-06 L07003-03
AAAL07003-06EA 269.99 USD InStock
AAAL07003-06 CAAAAL07003-06 AAAL07003-03 CAAAAL07003-03
2-Amino-3,5-dimethylbenzoic acid 98%
2-Amino-3,5-dimethylbenzoic acid


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00017099
CAS Number: 14438-32-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty