Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluorobenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Bromo-2-fluorophenyl)methanol4-Bromo-2-fluorobenzyl Alcohol

L18679-03 L18679-06 L18679-06 L18679-03
AAAL18679-03EA 39.28 USD InStock
CAAAAL18679-03 AAAL18679-03 CAAAAL18679-06 AAAL18679-06
4-Bromo-2-fluorobenzyl alcohol 97%
4-Bromo-2-fluorobenzyl alcohol


Formula: C₇H₆BrFO
MW: 205.03 g/mol
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00143265
CAS Number: 188582-62-9
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty