Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Trifluoromethoxy)thiophenol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Trifluoromethoxy)benzene-1-thiol2-(Trifluoromethoxy)benzenethiol

L19911-03 L19911-06 L19911-03 L19911-06
AAAL19911-06EA 397.99 USD InStock
AAAL19911-06 CAAAAL19911-03 CAAAAL19911-06 AAAL19911-03
2-(Trifluoromethoxy)thiophenol 99%
2-(Trifluoromethoxy)thiophenol


Formula: C₇H₅F₃OS
MW: 194.18 g/mol
Boiling Pt: 53…55 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00052320
CAS Number: 175278-01-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty