Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Bromo-2,3,5,6-tetrafluorobenzene-1-carbonyl chloride

L20153-14 L20153-03 L20153-03 L20153-14 L20153-06 L20153-06
AAAL20153-03EA 71.07 USD InStock
CAAAAL20153-14 CAAAAL20153-03 CAAAAL20153-06 AAAL20153-03 AAAL20153-14 AAAL20153-06
4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride 98%
4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride


Formula: C₇BrClF₄O
MW: 291.43 g/mol
Boiling Pt: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD03001173
CAS Number: 122033-54-9
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty