Would you like to log in again?
Drosophila melanogaster GenLysate™ Insect Lysate, G-Biosciences
Supplier: G-Biosciences
ML-01 ML-01
82021-660EA 180.11 USD InStock
82021-660 CA82021-660
Drosophila melanogaster GenLysate™ Insect Lysate, G-Biosciences
Cells
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes
G-Biosciences' GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris·HCl and 2% SDS with G-Biosciences' ProteaseARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty