Would you like to log in again?
Safety-Coated Straight Sided Round Bottles, Clear, Wide Mouth
Supplier: Qorpak
GLA-00946 GLC-02242
16195-113CS 298.16 USD InStock
16195-113 76539-352
Safety-Coated Straight Sided Round Bottles, Clear, Wide Mouth
Bottles Glass Bottles
Reduce breakage and lab mishaps with these safety coated clear straight sided round bottles.
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty