Would you like to log in again?
Bottles, Cylinder, White High-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PLA-05755 PLA-05753 PLA-05751
89204-908CS 150.18 USD InStock
89204-908 16087-408 16087-404
Bottles, Cylinder, White High-Density Polyethylene, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
White HDPE cylinder bottles are impact resistant.
They provide an excellent moisture barrier and offer a degree of light protection. Ideal for pharmaceutical applications. Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty