Would you like to log in again?
Packer Bottles, PET, Amber, Wide Mouth, Qorpak
Supplier: Qorpak
PLC-06593 PLC-06592
11311-372CS 121.27 USD InStock
11311-372 11311-370
Packer Bottles, PET, Amber, Wide Mouth, Qorpak
Bottles Roller Bottles
PET Packer Bottles feature glass like clarity and are durable and rigid.
  • PET packers are stress and impact resistant
  • PET bottles are ideal for oil analysis
They have exceptional gas and moisture barrier properties and a good alcohol and solvent barrier.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty