Would you like to log in again?
Nalgene® Diagnostic Bottles, Natural PP, with Closures, Tray Packed, Thermo Scientific
Supplier: Thermo Scientific
322006-9050 322006-9025 322006-9125
16059-690CS 1436.54 USD InStock
16059-690 16059-698 16059-682
Nalgene® Diagnostic Bottles, Natural PP, with Closures, Tray Packed, Thermo Scientific
Bottles Plastic Bottles
Nalgene natural polypropylene diagnostic bottles with natural polypropylene closures are used in test kits and many small volume applications.
Diagnostic bottles can be used for sample storage and shipping of small volume, valuable reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty