Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
White Polypropylene Hole Caps, Bonded PTFE/Silicone Septa, Qorpak®
Supplier: Qorpak
CAP-00355 CAP-00343
16200-422CS 3216.1 USD InStock
16200-422 16151-387
White Polypropylene Hole Caps, Bonded PTFE/Silicone Septa, Qorpak®
Closures Bottle Closures
White Polypropylene Hole Caps with bonded PTFE/Silicone Septa provide assurance that the septa will not slip away from the cap while injecting
The PTFE/Silicone septa has excellent resealing and chemical resistant properties.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty