Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM Fc5µ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
609-4131 609-4131
RL609-4131EA 189 USD InStock
RL609-4131 CA609-4131
Anti-IgM Fc5µ Rabbit Polyclonal Antibody
Antibodies
Human IgM Fc5µ antibody is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:40000 - 1:160,000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:500 - 1:3000;

Type: Secondary
Antigen: IgM Fc5µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Fc5µ
Host: Rabbit
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty