Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APOA1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-196 600-101-196
RL600-101-196EA 1351 USD InStock
RL600-101-196 CA600-101-196
Anti-APOA1 Goat Polyclonal Antibody
Antibodies
Anti-apoLipoProtein antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting for highly sensitive qualitative analysis.

Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunochemistry: 1:250 - 1:500; Immunoprecipitation: 1:100;

Type: Primary
Antigen: APOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty