Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
610-4120 610-4120
RL-610-4120EA 195 USD InStock
RL-610-4120 CA610-4120
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Antibody Anti-Mouse IgG (H&L) is suitable for immunoblotting (western or dot blot), ELISA, and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000 - 1:200000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty