Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-261 600-401-261
RL600-401-261EA 530 USD InStock
RL600-401-261 CARL600401-261
Anti-S Rabbit Polyclonal Antibody
Antibodies
Phosphoserine Antibody is suitable for use in ELISA, western blotting and immunoprecipitation assays.  The antibody reacts specifically with phosphoserine and shows minimal reactivity by ELISA and western blotting with Proteins containing phosphotyrosine.

Recommended Dilutions: ELISA: 1:1000-1:2500; Western Blot: 1:500-1:1000; Immunoprecipitation: 10 µg/mg;

Type: Primary
Antigen: S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty