Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AAO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-4148 100-4148
RL100-4148EA 1289.92 USD InStock
RL100-4148 CA100-4148
Anti-AAO Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-ASCORBATE OXIDASE (cucurbita species) Reacts with Pan

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: AAO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cucurbita

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty