Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-161 100-401-161
RL100-401-161EA 518 USD InStock
RL100-401-161 CA100-401-161
Anti-CDK2 Rabbit Polyclonal Antibody
Antibodies
Anti-cdk2 antbody is suitable for immunochemistry, western blotting and other antibody based assays.  Multiple isoforms as well as phosphorylated forms of cdk-2 may be detected by this antibody. Use paraffin embedded tissues for immunochemistry.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty