Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIVM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-250 100-401-250
RL100-401-250EA 518 USD InStock
RL100-401-250 CA100-401-250
Anti-BIVM Rabbit Polyclonal Antibody
Antibodies
This polyclonal antibody reacts human BIVM at the C-terminal region and is useful for detection of BIVM by ELISA or western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000;

Type: Primary
Antigen: BIVM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty