Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MIF Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-457 200-901-457
RL200-901-457EA 570 USD InStock
CA200-901-457 RL200-901-457
Anti-MIF Chicken Polyclonal Antibody
Antibodies
This IgY fraction antibody of anti-Human macrophage migration inhibitory factor (MIF) has been tested for use in ELISA and immunoblotting.  Although not tested, this antibody may also be useful for neutralization assays.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 500 µl

Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty