Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDKN2A Rat Polyclonal Antibody [clone: 5.C3.1]

Supplier: Rockland Immunochemicals
200-501-891 200-501-891
RL200-501-891EA 528 USD InStock
CA200-501-891 RL200-501-891
Anti-CDKN2A Rat Polyclonal Antibody [clone: 5.C3.1]
Antibodies
This antibody has been tested for use in ELISA, western blotting, immunofluorescence microscopy, and immunoprecipitation. Specific conditions for western blotting reactivity should be optimized by the end user.  Expect a band approximately 19 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Immunofluorescence: 1:500 - 1:2000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone: 5.C3.1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty