Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIK3CD Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-862 100-401-862
RL100-401-862EA 518 USD InStock
RL100-401-862 CA100-401-862
Anti-PIK3CD Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA, western blotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.  A mouse whole cell splenic lysate is suitable for use as a positive control.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:5000;

Type: Primary
Antigen: PIK3CD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty