Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
611-1623 611-1623
RL611-1623EA 269 USD InStock
RL611-1623 CA611-1623
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-RABBIT IgG F(c) (GOAT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoelectron microscopy and immunochemistry as well as other antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:115000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty