Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Methyl Lysine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
600-403-940 600-403-940
RL600-403-940EA 539 USD InStock
CARL600403-940 RL600-403-940
Anti-Methyl Lysine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
This affinity-purified antibody is suitable for use in ELISA and western blotting.  The antibody reacts specifically with methylated lysine residues.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000-1:5000;

Type: Primary
Antigen: Methyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Mammalian

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty