Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

600-100-215 600-100-215
RL600-100-215EA 686 USD InStock
CA600-100-215 RL600-100-215
Anti-GFP Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Primary Goat Anti-GFP Min X Hu Ms and Rt Serum Proteins Reacts with Pan

Recommended Dilutions: Immunofluorescence: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty