Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, Proteomics Grade

Supplier: G-Biosciences
Synonyms: Dihydrogen oxide

786-229 786-229
82021-370EA 59.86 USD InStock
82021-370 CA82021-370
Water, Proteomics Grade
Water
G-Biosciences' Proteomic-Grade Water is suitable for 2D electrophoresis and mass spectrometry analysis. Removes worries of protien and dust contamination and improves quality and reproducibility of 2D electrophoresis and mass spectrometry results.

G-Biosciences also offers Endotoxin-Free, Molecular-Grade, and DEPC-Treated Water.


Formula: H₂O
MW: 18.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty