Would you like to log in again?
Affinity Purified Avidin, G-Biosciences
Supplier: G-Biosciences
786-583 786-581 786-583 786-582 786-581 786-582
71003-198EA 867.16 USD InStock
71003-198 CA71003-196 CA71003-198 71003-194 CA71003-194 71003-196
Affinity Purified Avidin, G-Biosciences
Protein Linking and Labeling Systems
G-Biosciences' avidin is a glycoprotein with approximately 10% of its total mass comes from carbohydrates
  • Specific activity 15.3 U/mg protein
Avidin is a basic protein with an isoelectric pH of 10 to 10.5 and is readily soluble in aqueous buffers containing a wide range of salt, pH, temperature and other laboratory agents. This wide range of tolerance makes avidin suitable for a wide variety of analytical applications.
G-Biosciences' affinity purified avidin is ideal for estimation of biotin incorporation and other applications.
Avidin has a molecular weight of 67 kDa and contains four identical 128 amino acid subunits that each have a single biotin binding domain.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty