Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mineral oil, white (petroleum)

Supplier: G-Biosciences
Warning
Synonyms: Matrix oilMineral oilParaffin oil

RC-118 RC-118
71003-510EA 103.48 USD InStock
71003-510 CA71003-510
Mineral oil, white (petroleum)
White mineral oil (petroleum)


Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty