Would you like to log in again?
BioSpec-nano Small-volume UV Spectrophotometer, Shimadzu
Supplier: Shimadzu
2062630042
89210-930EA 20579.45 USD InStock
89210-930
BioSpec-nano Small-volume UV Spectrophotometer, Shimadzu
Spectrophotometers and Photometers UV/Visible Spectrophotometers
A state-of-the-art UV-VIS Spectrophotometer for life sciences, the small-volume, completely automated BioSpec-nano features an outstanding analysis range and excellent measurement reproducibility, making it the solution for stress-free nucleic acid quantitation and protein analysis.
Key features include an ultra-small sample volumes (1μL - 2μL), automatic sample mounting, measurement and cleaning, and easy operation with "Drop-and-Click" analysis. In addition, the BioSpec-nano ensures excellent measurement reproducibility, enables photometric analysis (up to 8 wavelengths), and can be equipped with an optional 5mm pathlength cell.
Certifications: UL Listed.
Ordering information: Accessories not included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty