Would you like to log in again?
Precoiled Pipe Markers- 'Air B/Y', National Marker
Supplier: National Marker
B4003 H4003 E4003 G4003 C4003 D4003 A4003 F4003
94014-290EA 64.41 USD InStock
94014-290 94015-070 94009-352 94013-744 94004-060 94005-436 94012-548 94003-286
Precoiled Pipe Markers- 'Air B/Y', National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty