Would you like to log in again?
Cold Finger, Condenser, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
9720-23 9720-13
89075-462EA 74.81 USD InStock
89075-462 89075-460
Cold Finger, Condenser, Microscale, Ace Glass Incorporated
Condensers
Microscale cold finger condenser with outside body diameter to fit two sizes of Ace-Thred bushings
Water connection tubing is 5mm O.D. Overall body length is approximately 200mm. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty