Would you like to log in again?
Instatherm® Heated Filter Reactor Body, Ace Glass Incorporated
Supplier: Ace Glass
6388-15 6388-30 6388-35 6388-20 6388-40 6388-25
89054-170EA 4078.98 USD InStock
89054-170 89054-160 89054-162 89054-164 89054-166 89054-168
Instatherm® Heated Filter Reactor Body, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Rugged, heavy-wall reactor flask, Instatherm® coated for safer, rapid heating. Flange at top is Duran style 60 mm (2.4-inch), 100 mm (4-inch), or 150 mm (6-inch) with shallow o-ring groove, for connection various heads with quick release clamp. Bottom has a #50 or #80 Ace-Thred (internal glass thread) for installing bottom adapter with filter support and drain valve.
Instatherm® is an integral noble metal alloy fused permanently to the glass and covered with a tough silicone rubber-treated glass cloth insulation which serves as a thermal barrier as well as protection against physical shock. Heat response is rapid and thermal lag low typical heating rates are approximately 5° per minute. Instatherm® can be operated at 120v or 40v with ACE or J-Kem temperature controllers.
Can be used with both regular and pressure version heads and accessories. Fabricated from borosilicate glass.
Flange at top is Duran style 60 mm (2.4-inch), 100 mm (4-inch), or 150 mm (6-inch) with shallow o-ring groove, for connection various heads with quick release clamp. Bottom has a #50 or #80 Ace-Thred (internal glass thread) for installing bottom adapter with filter support and drain valve.
Ordering information: Head, bottom adapter, and other accessory items, order separately. Supplied with cord and CAPFE o-ring. Maximum temperature (with no vacuum or pressure) is 200°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty