Would you like to log in again?
Precoiled Pipe Markers- 'Fuel Oil Return' B/Y, National Marker
Supplier: National Marker
C4047 G4047 E4047 B4047 A4047 F4047 D4047 H4047
94015-158EA 87.99 USD InStock
94015-158 94013-832 94009-484 94012-636 94003-374 94014-378 94005-564 94004-148
Precoiled Pipe Markers- 'Fuel Oil Return' B/Y, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty