Would you like to log in again?
Molecular BioProducts Solvent Safe Carbon Filtered Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
5069B 5079EB
72830-014CS 1156.04 USD InStock
72830-014 72830-004
Molecular BioProducts Solvent Safe Carbon Filtered Pipette Tips, Thermo Scientific
Pipette Tips
MicroPoint design for accurate low-volume pipetting.
  • Volume: 100 to 1000 µl
  • Non-sterile
  • Certified RNase-, DNase-, DNA- and pyrogen-free
  • Bulk pack
Tips, pipette, filter, 200 µl, MicroPoint, with reference marks, Manual pipetting of organic solvents, 500 tips per bag.
Solvent safe carbon filter protects pipettors from damage by blocking corrosive vapors from volatile organic solvents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty