Would you like to log in again?
Precoiled Pipe Markers- 'Overflow', Black/Yellow, National Marker
Supplier: National Marker
A4078 B4078 F4078 D4078 G4078 E4078 C4078 H4078
94004-210EA 20.72 USD InStock
94004-210 94003-436 94005-650 94009-582 94012-698 94013-894 94015-220 94014-440
Precoiled Pipe Markers- 'Overflow', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty