Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Precoiled Pipe Markers- 'Domestic Hot Water Return', Black/Yellow, National Marker
Supplier: National Marker
D4037 C4037 G4037 F4037 B4037 E4037 A4037 H4037
94009-456EA 40.82 USD InStock
94009-456 94015-138 94014-358 94013-812 94004-128 94012-616 94003-354 94005-534
Precoiled Pipe Markers- 'Domestic Hot Water Return', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty