Would you like to log in again?
Precoiled Pipe Markers- 'Exhaust', Black/Yellow, National Marker
Supplier: National Marker
C4041 F4041 D4041 E4041 H4041 A4041 B4041 G4041
94005-552EA 41.82 USD InStock
94005-552 94013-820 94009-472 94012-624 94014-366 94003-362 94015-146 94004-136
Precoiled Pipe Markers- 'Exhaust', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty