Would you like to log in again?
Precoiled Pipe Markers 'Industrial Water', White/Green, National Marker
Supplier: National Marker
B4065 A4065 F4065 D4065 E4065 C4065 H4065 G4065
94013-868EA 46.11 USD InStock
94013-868 94012-672 94004-184 94014-414 94015-194 94005-614 94009-548 94003-410
Precoiled Pipe Markers 'Industrial Water', White/Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty