Would you like to log in again?
Precoiled Pipe Markers 'Low Pressure Steam', Black/Yellow, National Marker
Supplier: National Marker
A4069 F4069 H4069 C4069 B4069 D4069 G4069 E4069
94009-556EA 42.82 USD InStock
94009-556 94015-202 94003-418 94013-876 94004-192 94014-422 94005-622 94012-680
Precoiled Pipe Markers 'Low Pressure Steam', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty