Would you like to log in again?
Precoiled Pipe Markers 'Boiler Feed', Black/Yellow, National Marker
Supplier: National Marker
D4008 C4008 B4008 F4008 G4008 E4008 H4008 A4008
94009-362EA 42.82 USD InStock
94009-362 94013-754 94015-080 94012-558 94004-070 94014-300 94003-296 94005-446
Precoiled Pipe Markers 'Boiler Feed', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty