Would you like to log in again?
Precoiled Pipe Markers 'Chlorine', Black/Yellow, National Marker
Supplier: National Marker
C4016 B4016 F4016 D4016 A4016 G4016 H4016 E4016
94005-472EA 41.82 USD InStock
94005-472 94012-574 94009-386 94004-086 94014-316 94015-096 94013-770 94003-312
Precoiled Pipe Markers 'Chlorine', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty