Would you like to log in again?
Precoiled Pipe Markers 'Condenser Water', White/Green, National Marker
Supplier: National Marker
H4028 C4028 E4028 B4028 A4028 D4028 G4028 F4028
94003-336EA 16.1 USD InStock
94003-336 94004-110 94005-506 94012-598 94015-120 94009-418 94013-794 94014-340
Precoiled Pipe Markers 'Condenser Water', White/Green, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty