Would you like to log in again?
Precoiled Pipe Markers 'Return', Black/Yellow, National Marker
Supplier: National Marker
G4092 H4092 D4092 A4092 B4092 C4092 E4092 F4092
94015-248EA 87.99 USD InStock
94015-248 94014-468 94013-922 94003-464 94004-238 94012-726 94009-626 94005-696
Precoiled Pipe Markers 'Return', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty