Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Precoiled Pipe Markers 'Vent', Black/Yellow, National Marker
Supplier: National Marker
F4109 H4109 D4109 B4109 E4109 C4109 G4109 A4109
94013-956EA 46.11 USD InStock
94013-956 94012-760 94004-272 94014-502 94015-282 94005-736 94009-678 94003-498
Precoiled Pipe Markers 'Vent', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certifications: Meets ANSI A13.1-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty