Would you like to log in again?
Nalgene® End Caps, Thermo Scientific
Supplier: Thermo Scientific
2689-2075 2688-2075
73520-512CS 103.55 USD InStock
73520-512 73520-510
Nalgene® End Caps, Thermo Scientific
Fittings and Connectors
Thermo Scientific Nalgene End Caps for Carboys in PC (polycarbonate) or PP (polypropylene) provide sanitary ports, which allow easy fill/dispense operations.
Securely mounts to 3 inch sanitary flange for applications used with Nalgene Sanitary Carboys. Compatible with Nalgene Carboy 2630 and 2261.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty