Would you like to log in again?
Linen Hampers, Lakeside®
Supplier: Lakeside
4515 4518
89210-238EA 1448.05 USD InStock
89210-238 89210-240
Linen Hampers, Lakeside®
Carts
Open construction designed for easy unloading and minimal lifting
Grey-epoxy coated steel lid designed with steel lid prop to stay open when needed. Built in bag holder provides heavy duty support for 36 or 42 gallon bags. Foot operated lid mechanism allows sanitary hands-free operation. 3" locking rubber wheel ball bearing casters for smooth movement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty