Would you like to log in again?
Sign Adapter, National Marker
Supplier: National Marker
HDSAW HDSAY
94015-622EA 19.11 USD InStock
94015-622 94015-620
Sign Adapter, National Marker
Signs
Fits in top of warning posts
Accommodates signs up to .48cm (3/16") thick. Adapter Size: 2.54cm x 13.97cm (1" x 5½"). Signs and posts sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty