Would you like to log in again?
Tweezer, Bent General Purpose, Serrated, Excelta Corp®
Supplier: Excelta
24A-S-SE 20A-1-SE 24-8-SA-SE 24-SA-PI 24-6-SA 24-SA-SE
63041-634EA 11.33 USD InStock
63041-634 63041-638 89411-734 75880-818 63041-620 63041-640
Tweezer, Bent General Purpose, Serrated, Excelta Corp®
Forceps
These are great tools for general assembly (large parts) and feature serrated tips and handles for positive gripping.
  • Serrated grips for better control
  • Serrated tips for gripping power
  • Cleanroom safe, stainless steel construction
These 45° bent strong serrated tipped tweezer are ideal for general applications. They are made of anti-magnetic stainless steel and have serrated tips and grips for better control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty