Would you like to log in again?
VWR® Serum Vials / Diagnostic Bottles, PETG
Supplier: VWR
VWR®
RGC0010S RGC0005S
89207-574CS 1671.92 USD InStock
89207-574 89207-572
VWR® Serum Vials / Diagnostic Bottles, PETG
Vials
All VWR® sterile labware, media bottles, vials, and culture vials are manufactured with the remarkable Eastar PETG resin from Eastman.
  • Sterile
  • Excellent clarity
  • Tamper-evident design
VWR® products retain their clarity after gamma sterilization, maintain integrity of the contents, and more, so they are guaranteed to exceed customers’ expectations and requirement.
VWR® polythylene terephthalate copolyester (PETG) sterile serum vials are ideal for diagnostic specimen, safe storage, transfer and transport of antibodies, proteins, nucleic acids, enzyme and substrates. All VWR® PETG vials meet with current biological and pharmaceutical regulatory standards, such as: USP Class VI, non-cytotoxic, non-pyrogenic, USP <661>, European Pharmacopeia for Abnormal toxicity. They are made from ADCF (Animal Derived Component Free) materials. PETG vials are suitable for air shipment and are tested during each manufacturing run to ensure leak-proof performance.
Our line of tamper-evident media bottles and vials are exclusively made with this resin to satisfy the critical requirements of the in-vitro fertilization media, and all cell culture fluids.
It is an amorphous material that displays performance properties including clarity, toughness, excellent gas barrier and chemical inertness.The products can be safely sterilized with gamma radiation without property loss or color shift.
Certifications: VWR® PETG resin meets ISO 10993 and/or USP Class VI biocompatibility requirement; Food Contact Status compliant.
Ordering information: They are packaged with a blue tamper-evident, polyethylene closures. All vials are provided sterile SAL10⁻⁶, in a multi-barrier, cleanroom production packaging with caps engaged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty