Would you like to log in again?
Adapter, Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5218-10
89082-642EA 45.12 USD InStock
89082-642
Adapter, Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features an o-ring joint at one end and a hose connection at the other end.
  • O-ring joint
  • With hose connection
  • Fabricated from borosilicate glass
Adapter has a #15 o-ring joint at one end and a hose connection at the other end. Use with a 3/8-inch or 7/16-inch I.D. tubing, size E hose connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty