Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
AdaptecTM Series Protective Eyewear, Honeywell
Supplier: Honeywell Safety
Adaptec
T5900LTK T5900LTK T5900NTKS T5900LTKS T5900WTKS T5900NTKS T5900NTK T5900LTKS T5900NTK T5900WTK
89204-610BX 343.27 USD InStock
89204-610 CA89204-610 89204-622 CA89204-594 89204-594 CA89204-606 CA89204-590 89204-606 89204-590 CA89204-626
AdaptecTM Series Protective Eyewear, Honeywell
Safety Glasses
Adjustable eyewear system is designed for a customized fit while still delivering excellent protection, superior comfort, premium optics, and a stylish look.
  • Three-position adjustable nose bridge keeps eyes centered in lens
  • Self-adjusting nose pads independently conform to the wearer’s face for greater comfort and stability
  • Temples adjust for inclination and width without removing the eyewear
  • Quick-change lens replacement system
Available in three sizes to provide the right protection, improved performance, and optical clarity for everyone in your workplace. Utilizes exclusive Adaptec™ 4A+ coating, protecting against fog, scratches, UV, and static. Translucent black frame color.
Certifications: Meets ANSI Z87.1 and CSA Z94.3 requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty