Would you like to log in again?
New Brunswick Excella Incubator Shaker E25 / E25R, Eppendorf
Supplier: Eppendorf
M1353-0004 M1353-0004 M1353-0000 M1353-0000
89171-828EA 24924.93 USD InStock
CA89171-828 89171-828 CA89171-824 89171-824
New Brunswick Excella Incubator Shaker E25 / E25R, Eppendorf
Shakers and Mixers Shaking Incubators
The New Brunswick Excella model E25 and refrigerated E25R biological shakers provide advanced features and excellent performance at an affordable price
A large 30" x 18" accessory platform (sold separately) accommodates flasks up to 6 L with a large selection of interchangeable platforms and varying clamp sizes. The wide temperature range of the E25: 7 °C above ambient to 60 °C. (E25R: 15 °C below ambient to 60 °C) is supported by powerful fans and heater to provide rapid heat up and temperature recovery after door opening. Audible and visual alarms alert you to set point deviations or end of study. RS -232 interface for data logging directly to your PC
The large capacity console style incubator with top-loading door allows easy viewing of your culture media through a double-paned, insulated window. This model features the New Brunswick heavy-duty counterbalanced shaking mechanism for uniform motion and years of quiet, dependable operation. Microprocessor controls with digital display enable easy operation and accurate set point regulation of temperature, speed and running time. A large assortment of platforms, clamps, test tube holders and other accessories allows configuration to meet your exact needs.
Certifications: cETLus Certified: meets CSA, UL and CE standards
Ordering information: New Brunswick Excella Series shakers come with a two-year manufacturer’s warranty on both parts and labor. All platforms, accessory clamps and racks sold separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty